Gene Transcripts

Gene Symbol
CARD16
Entrez Gene ID
114769
Gene Name
caspase recruitment domain family member 16
Gene Aliases
COP, COP1, PSEUDO-ICE
Chromosome Location
Chr. 11: 105040218 - 105045416 on Build GRCh38
UniGene ID
Hs.348365
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001017534.1 NP_001017534.1 3 424
NM_052889.2 NP_443121.1 4 487
XM_011542583.2 XP_011540885.1 3 616
GenBank mRNA AK125640.1 - 2 3008
AY885669.1 - 3 415
BC070196.1 - 4 487
BC104635.1 - 4 491

View Full Product Details