Gene Transcripts

Gene Symbol
TM4SF19
Entrez Gene ID
116211
Gene Name
transmembrane 4 L six family member 19
Gene Aliases
OCTM4
Chromosome Location
Chr. 3: 196323546 - 196338420 on Build GRCh38
UniGene ID
Hs.733264
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001204898.1 NP_001191827.1 3, 4 522
NM_138461.3 NP_612470.2 4, 5 600
GenBank mRNA AF192910.1 - 4, 5 556
BC146985.1 - 4, 5 501 9056913
BC146993.1 - 4, 5 501 9056921
BC147021.1 - 4, 5 501 9056949
BC171870.1 - 3, 4 423 9054524
BP395549.1 - 1, 2 40
BQ678650.1 - 4, 5 566
CA430273.1 - 1, 2 474
CA489370.1 - 3, 4 522

Gene Symbol
TM4SF19-TCTEX1D2
Entrez Gene ID
100534611
Gene Name
TM4SF19-TCTEX1D2 readthrough (NMD candidate)
Gene Aliases
Chromosome Location
Chr. 3: 196316085 - 196338420 on Build GRCh38
UniGene ID
Hs.733180
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_037950.1 4, 5 600

View Full Product Details