Gene Transcripts
- Gene Symbol
- JAML
- Entrez Gene ID
- 120425
- Gene Name
- junction adhesion molecule like
- Gene Aliases
- AMICA, AMICA1, CREA7-1, CREA7-4, Gm638
- Chromosome Location
- Chr. 11: 118193727 - 118225094 on Build GRCh38
- UniGene ID
- Hs.16291
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001098526.1 |
NP_001091996.1
|
3, 4 |
378 |
|
NM_001286570.1 |
NP_001273499.1
|
2, 3 |
315 |
|
NM_001286571.1 |
NP_001273500.1
|
2, 3 |
606 |
|
NM_153206.2 |
NP_694938.2
|
2, 3 |
616 |
|
NR_104479.1 |
|
2, 3 |
606 |
|
XM_005271407.3 |
XP_005271464.1
|
2, 3 |
316 |
|
XM_017017213.1 |
XP_016872702.1
|
3, 4 |
303 |
|
XM_017017214.1 |
XP_016872703.1
|
3, 4 |
373 |
|
XM_017017215.1 |
XP_016872704.1
|
2, 3 |
301 |
|
GenBank mRNA |
AJ515553.2 |
- |
3, 4 |
298 |
|
AK090409.1 |
- |
3, 4 |
278 |
|
AK094399.1 |
- |
2, 3 |
616 |
|
AK298722.1 |
- |
2, 3 |
232 |
|
AK300851.1 |
- |
3, 4 |
295 |
|
AK301099.1 |
- |
3, 4 |
295 |
|
AY093686.1 |
- |
3, 4 |
295 |
|
AY093687.1 |
- |
1, 2 |
88 |
|
AY138965.1 |
- |
3, 4 |
279 |
|
AY358362.1 |
- |
3, 4 |
288 |
|
BC100794.2 |
- |
2, 3 |
305 |
40000649 |
BC100795.2 |
- |
2, 3 |
305 |
40000652 |
BC100796.1 |
- |
2, 3 |
305 |
40000654 |
BC100797.1 |
- |
2, 3 |
305 |
40000656 |
DA090367.1 |
- |
2, 3 |
328 |
|
DC296115.1 |
- |
3, 4 |
378 |
|
View Full Product Details