Gene Transcripts

Gene Symbol
MYO3B
Entrez Gene ID
140469
Gene Name
myosin IIIB
Gene Aliases
Chromosome Location
Chr. 2: 170178145 - 170655325 on Build GRCh38
UniGene ID
Hs.671900
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001083615.3 NP_001077084.2 9 1007
NM_138995.4 NP_620482.3 9 1007
NR_045682.1 9 1007
NR_045683.1 9 1007
NR_045684.1 9 1007
XM_006712299.3 XP_006712362.1 9 895
XM_011510654.2 XP_011508956.1 9 895
XM_011510655.2 XP_011508957.1 9 895
XM_011510656.2 XP_011508958.1 2 126
XM_011510657.2 XP_011508959.1 2 142
XM_011510658.2 XP_011508960.1 2 307
GenBank mRNA AF369908.1 - 9 1007
AF391554.1 - 9 1007
AF391555.1 - 9 1007
AF391556.1 - 9 1007
AF391557.1 - 9 1007
AF391558.1 - 9 1007
AK055778.1 - 9 895
BC040019.1 - 8 862
BC136620.1 - 9 944 9020610
BC144307.1 - 9 944 9052827
BX647895.1 - 7 732

View Full Product Details