Gene Transcripts

Gene Symbol
RNF32
Entrez Gene ID
140545
Gene Name
ring finger protein 32
Gene Aliases
FKSG33, HSD15, LMBR2
Chromosome Location
Chr. 7: 156640286 - 156678333 on Build GRCh38
UniGene ID
Hs.446194
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001184996.1 NP_001171925.1 9 1410
NM_001184997.1 NP_001171926.1 9 1263
NM_001308273.1 NP_001295202.1 9 1485
NM_030936.3 NP_112198.1 9 1305
NR_131778.1 9 1259
XM_005249522.4 XP_005249579.1 9 1217
XM_011515804.2 XP_011514106.1 10 1327
XM_011515805.2 XP_011514107.1 10 1484
XM_011515806.2 XP_011514108.1 9 1395
XM_011515807.2 XP_011514109.1 9 1203
XM_011515808.2 XP_011514110.1 9 1315
XM_011515809.2 XP_011514111.1 9 1388
XM_011515810.2 XP_011514112.1 9 1205
XM_011515811.2 XP_011514113.1 9 1200
XM_011515812.2 XP_011514114.1 9 1307
XM_011515813.2 XP_011514115.1 9 1266
GenBank mRNA AF325690.1 - 9 1213
AF441222.1 - 8 1152
AF441224.1 - 9 1259
AK314067.1 - 9 1411
AL136874.1 - 9 1193
CR533513.1 - 7 1075

View Full Product Details