Gene Transcripts

Gene Symbol
NKX2-5
Entrez Gene ID
1482
Gene Name
NK2 homeobox 5
Gene Aliases
CHNG5, CSX, CSX1, HLHS2, NKX2.5, NKX2E, NKX4-1, VSD3
Chromosome Location
Chr. 5: 173232104 - 173235312 on Build GRCh38
UniGene ID
Hs.54473
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001166175.1 NP_001159647.1 2 1469
NM_001166176.1 NP_001159648.1 2 1432
NM_004387.3 NP_004378.1 2 1177
GenBank mRNA AB021133.1 - 2 948
AI609745.1 - 1 489
AK290615.1 - 2 1069
BC025711.1 - 2 1055 5225103
KU178001.1 - 2 948
KU178002.1 - 2 1240
U34962.1 - 2 1124

View Full Product Details