Gene Transcripts
- Gene Symbol
- FBXO41
- Entrez Gene ID
- 150726
- Gene Name
- F-box protein 41
- Gene Aliases
- FBX41
- Chromosome Location
- Chr. 2: 73254682 - 73284453 on Build GRCh38
- UniGene ID
- Hs.23158
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001080410.2 |
NP_001073879.2
|
10, 11 |
2454 |
|
XM_005264158.3 |
XP_005264215.1
|
8, 9 |
1373 |
|
XM_006711950.2 |
XP_006712013.1
|
15, 16 |
3438 |
|
XM_011532562.1 |
XP_011530864.1
|
11, 12 |
2886 |
|
XM_011532563.2 |
XP_011530865.1
|
9, 10 |
1811 |
|
XM_017003421.1 |
XP_016858910.1
|
12, 13 |
5535 |
|
XM_017003422.1 |
XP_016858911.1
|
15, 16 |
5180 |
|
XM_017003423.1 |
XP_016858912.1
|
15, 16 |
4561 |
|
GenBank mRNA |
AB075820.1 |
- |
10, 11 |
2292 |
|
AK095028.1 |
- |
1, 2 |
125 |
|
AK127503.1 |
- |
7, 8 |
935 |
|
BC112202.1 |
- |
9, 10 |
1362 |
|
View Full Product Details