Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001321498.1 | NP_001308427.1 | 4 | 766 | |
NM_001321500.1 | NP_001308429.1 | 3 | 697 | ||
NM_001321501.1 | NP_001308430.1 | 4 | 770 | ||
NM_001321502.1 | NP_001308431.1 | 3 | 697 | ||
NM_001321509.1 | NP_001308438.1 | 4 | 770 | ||
NM_001321512.1 | NP_001308441.1 | 4 | 766 | ||
NM_001321513.1 | NP_001308442.1 | 4 | 903 | ||
NM_001321514.1 | NP_001308443.1 | 4 | 594 | ||
NM_001321515.1 | NP_001308444.1 | 3 | 834 | ||
NM_144710.4 | NP_653311.1 | 4 | 766 | ||
NM_178584.3 | NP_848699.1 | 3 | 697 | ||
XM_006712317.1 | XP_006712380.1 | 4 | 766 | ||
XM_011510698.2 | XP_011509000.1 | 4 | 599 | ||
XM_011510699.2 | XP_011509001.1 | 3 | 418 | ||
XM_011510700.2 | XP_011509002.1 | 4 | 769 | ||
XM_011510703.2 | XP_011509005.1 | 4 | 376 | ||
XM_011510704.2 | XP_011509006.1 | 4 | 766 | ||
GenBank mRNA | AF146760.1 | - | 4 | 513 | |
AK021681.1 | - | 4 | 766 | ||
AK022682.1 | - | 1 | 98 | ||
AK092033.1 | - | 3 | 417 | ||
AK292038.1 | - | 4 | 484 | ||
BC020502.1 | - | 4 | 460 | 3913356 | |
BC050345.1 | - | 4 | 703 | 4830029 |