Gene Transcripts

Gene Symbol
NFXL1
Entrez Gene ID
152518
Gene Name
nuclear transcription factor, X-box binding like 1
Gene Aliases
CDZFP, HOZFP, OZFP, URCC5
Chromosome Location
Chr. 4: 47847233 - 47914667 on Build GRCh38
UniGene ID
Hs.646325
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001278623.1 NP_001265552.1 7 1008
NM_001278624.1 NP_001265553.1 7 1146
NM_152995.5 NP_694540.3 7 1048
NR_103795.1 7 1008
GenBank mRNA AB085695.1 - 7 1004
AB181916.1 - 7 1128
AK131285.1 - 5 594
AK302440.1 - 7 1006
BC051193.1 - 7 979 5271401

View Full Product Details