Gene Transcripts

Gene Symbol
NUDT17
Entrez Gene ID
200035
Gene Name
nudix hydrolase 17
Gene Aliases
Chromosome Location
Chr. 1: 145844985 - 145848954 on Build GRCh38
UniGene ID
Hs.585066
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012758.2 NP_001012776.1 6, 7 740
XM_011509260.2 XP_011507562.1 4, 5 1178
GenBank mRNA AA287243.1 - 1, 2 72
AK091752.1 - 2, 3 1999

View Full Product Details