Gene Transcripts

Gene Symbol
OR4C6
Entrez Gene ID
219432
Gene Name
olfactory receptor family 4 subfamily C member 6
Gene Aliases
OR11-138
Chromosome Location
Chr. 11: 55665167 - 55666096 on Build GRCh38
UniGene ID
Hs.511921
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004704.1 NP_001004704.1 1 893
GenBank mRNA BC137216.1 - 1 922
BC137218.1 - 1 922 9021215

View Full Product Details