Gene Transcripts

Gene Symbol
CHSY1
Entrez Gene ID
22856
Gene Name
chondroitin sulfate synthase 1
Gene Aliases
CHSY, CSS1, ChSy-1, TPBS
Chromosome Location
Chr. 15: 101175723 - 101251932 on Build GRCh38
UniGene ID
Hs.110488
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014918.4 NP_055733.2 3 2812
XM_006720435.3 XP_006720498.1 3 2094
XM_011521364.2 XP_011519666.1 4 2435
XM_017022011.1 XP_016877500.1 2 1868
GenBank mRNA AB023207.1 - 3 2830
AB071402.1 - 3 2350
AK075096.1 - 1 542
AK296943.1 - 2 1589
AY358529.1 - 3 2824
BC046247.1 - 3 2819 5751618

View Full Product Details