Gene Transcripts

Gene Symbol
P2RX2
Entrez Gene ID
22953
Gene Name
purinergic receptor P2X 2
Gene Aliases
DFNA41, P2X2
Chromosome Location
Chr. 12: 132618780 - 132623678 on Build GRCh38
UniGene ID
Hs.258580
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001282164.1 NP_001269093.1 9 947
NM_001282165.1 NP_001269094.1 10 1068
NM_012226.4 NP_036358.2 8 833
NM_016318.3 NP_057402.1 9 977
NM_170682.3 NP_733782.1 10 1049
NM_170683.3 NP_733783.1 10 1049
NM_174872.2 NP_777361.1 8 773
NM_174873.2 NP_777362.1 10 1049
XM_005266154.3 XP_005266211.1 9 1156
XM_005266155.4 XP_005266212.1 10 1043
XM_005266156.4 XP_005266213.1 10 1043
XM_011534786.2 XP_011533088.1 9 961
XM_017019035.1 XP_016874524.1 10 1043
GenBank mRNA AF109387.1 - 10 976
AF109388.1 - 10 976
AF190822.1 - 10 1012
AF190823.1 - 10 1012
AF190824.1 - 9 940
AF190825.1 - 10 1012
AF260426.1 - 10 1049
AF260427.1 - 10 1012
AF260428.1 - 8 736
AF260429.1 - 8 796
AY346374.1 - 9 910
BC109200.2 - 10 1031 40005772

View Full Product Details