Gene Transcripts

Gene Symbol
CLUAP1
Entrez Gene ID
23059
Gene Name
clusterin associated protein 1
Gene Aliases
CFAP22, FAP22, IFT38
Chromosome Location
Chr. 16: 3495562 - 3539048 on Build GRCh38
UniGene ID
Hs.155995
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015041.2 NP_055856.1 7 736
NM_024793.2 NP_079069.1 3 194
XM_006720867.3 XP_006720930.1 6 587
XM_017023068.1 XP_016878557.1 7 751
XM_017023069.1 XP_016878558.1 7 751
XM_017023070.1 XP_016878559.1 7 736
XM_017023071.1 XP_016878560.1 7 1725
XM_017023072.1 XP_016878561.1 7 1725
XM_017023073.1 XP_016878562.1 6 587
GenBank mRNA AB014543.1 - 7 663
AB089691.1 - 7 718
AK023359.1 - 3 194
BC017070.2 - 7 664 3916900

View Full Product Details