Gene Transcripts
- Gene Symbol
- FRMD4B
- Entrez Gene ID
- 23150
- Gene Name
- FERM domain containing 4B
- Gene Aliases
- 6030440G05Rik, GRSP1
- Chromosome Location
- Chr. 3: 69168782 - 69541340 on Build GRCh38
- UniGene ID
- Hs.371681
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_015123.2 |
NP_055938.1
|
20, 21 |
2353 |
|
XM_005264722.1 |
XP_005264779.1
|
20, 21 |
2030 |
|
XM_005264723.1 |
XP_005264780.1
|
10, 11 |
1512 |
|
XM_017005989.1 |
XP_016861478.1
|
20, 21 |
2137 |
|
XM_017005990.1 |
XP_016861479.1
|
20, 21 |
2030 |
|
XM_017005991.1 |
XP_016861480.1
|
21, 22 |
2190 |
|
XM_017005992.1 |
XP_016861481.1
|
20, 21 |
1970 |
|
XM_017005993.1 |
XP_016861482.1
|
20, 21 |
1961 |
|
XM_017005994.1 |
XP_016861483.1
|
20, 21 |
3803 |
|
XM_017005995.1 |
XP_016861484.1
|
14, 15 |
1547 |
|
XM_017005996.1 |
XP_016861485.1
|
10, 11 |
1249 |
|
XM_017005997.1 |
XP_016861486.1
|
10, 11 |
1512 |
|
GenBank mRNA |
AB023230.1 |
- |
20, 21 |
2122 |
|
AK024086.1 |
- |
4, 5 |
388 |
|
AK295045.1 |
- |
15, 16 |
1595 |
|
AL832231.1 |
- |
10, 11 |
1350 |
|
BC110637.1 |
- |
5, 6 |
574 |
|
View Full Product Details