Gene Transcripts

Gene Symbol
FAM189A1
Entrez Gene ID
23359
Gene Name
family with sequence similarity 189 member A1
Gene Aliases
TMEM228
Chromosome Location
Chr. 15: 29120252 - 29570723 on Build GRCh38
UniGene ID
Hs.383564
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015307.1 NP_056122.1 3 322
XM_011521407.2 XP_011519709.1 3 251
GenBank mRNA DA188047.1 - 3 318

View Full Product Details