Gene Transcripts
- Gene Symbol
- HARS2
- Entrez Gene ID
- 23438
- Gene Name
- histidyl-tRNA synthetase 2, mitochondrial
- Gene Aliases
- HARSL, HARSR, HO3, PRLTS2
- Chromosome Location
- Chr. 5: 140691426 - 140699318 on Build GRCh38
- UniGene ID
- Hs.432560
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001278731.1 |
NP_001265660.1
|
3, 4 |
542 |
|
NM_012208.3 |
NP_036340.1
|
4, 5 |
617 |
|
XM_011537619.1 |
XP_011535921.1
|
5, 6 |
630 |
|
XM_011537620.1 |
XP_011535922.1
|
5, 6 |
630 |
|
XM_017009287.1 |
XP_016864776.1
|
4, 5 |
617 |
|
XM_017009288.1 |
XP_016864777.1
|
5, 6 |
681 |
|
XM_017009289.1 |
XP_016864778.1
|
5, 6 |
674 |
|
XM_017009290.1 |
XP_016864779.1
|
4, 5 |
667 |
|
XM_017009291.1 |
XP_016864780.1
|
4, 5 |
360 |
|
XM_017009292.1 |
XP_016864781.1
|
4, 5 |
304 |
|
GenBank mRNA |
AK293390.1 |
- |
3, 4 |
359 |
|
AK309832.1 |
- |
4, 5 |
715 |
|
AK311277.1 |
- |
4, 5 |
713 |
|
AK312976.1 |
- |
4, 5 |
435 |
|
BC007680.2 |
- |
4, 5 |
436 |
3628006 |
BC014982.1 |
- |
4, 5 |
581 |
4859849 |
U18937.1 |
- |
4, 5 |
543 |
|
View Full Product Details