Gene Transcripts

Gene Symbol
RIMBP2
Entrez Gene ID
23504
Gene Name
RIMS binding protein 2
Gene Aliases
PPP1R133, RBP2, RIM-BP2
Chromosome Location
Chr. 12: 130396133 - 130716361 on Build GRCh38
UniGene ID
Hs.657441
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015347.4 NP_056162.4 14 2784
XM_011538102.1 XP_011536404.1 16 3602
XM_011538103.2 XP_011536405.1 18 4064
XM_011538104.2 XP_011536406.1 18 3879
XM_011538105.2 XP_011536407.1 17 3336
XM_011538106.2 XP_011536408.1 16 3177
XM_017019105.1 XP_016874594.1 18 4077
XM_017019106.1 XP_016874595.1 17 3336
XM_017019107.1 XP_016874596.1 17 3158
XM_017019108.1 XP_016874597.1 17 3341
XM_017019109.1 XP_016874598.1 15 2885
XM_017019110.1 XP_016874599.1 17 3158
XM_017019111.1 XP_016874600.1 17 3158
GenBank mRNA AB002316.1 - 14 2784
AY699979.1 - 11 2475
BX641152.1 - 1 7516

View Full Product Details