Gene Transcripts

Gene Symbol
VENTX
Entrez Gene ID
27287
Gene Name
VENT homeobox
Gene Aliases
HPX42B, NA88A, VENTX2
Chromosome Location
Chr. 10: 133237904 - 133241930 on Build GRCh38
UniGene ID
Hs.125231
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014468.3 NP_055283.1 2 341
XM_017016073.1 XP_016871562.1 2 336
GenBank mRNA AF068006.1 - 2 341
BC108915.1 - 2 341 40015551
BC108916.1 - 2 341 40015553

View Full Product Details