Gene Transcripts

Gene Symbol
BCL9L
Entrez Gene ID
283149
Gene Name
B-cell CLL/lymphoma 9-like
Gene Aliases
BCL9-2, DLNB11
Chromosome Location
Chr. 11: 118896140 - 118926072 on Build GRCh38
UniGene ID
Hs.414740
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_182557.2 NP_872363.1 6 3474
XM_005271511.2 XP_005271568.1 8 3474
XM_005271512.2 XP_005271569.1 7 2924
XM_005271513.2 XP_005271570.1 6 2923
XM_006718815.2 XP_006718878.1 7 2972
XM_011542756.2 XP_011541058.1 6 3430
XM_017017572.1 XP_016873061.1 8 3723
XM_017017573.1 XP_016873062.1 7 4075
XM_017017574.1 XP_016873063.1 7 3622
GenBank mRNA AB094091.1 - 6 3474
AK122882.1 - 1 1114
AY296059.1 - 6 2494
BC033257.1 - 1 393

View Full Product Details