Gene Transcripts

Gene Symbol
HLCS
Entrez Gene ID
3141
Gene Name
holocarboxylase synthetase
Gene Aliases
HCS
Chromosome Location
Chr. 21: 36750888 - 36990254 on Build GRCh38
UniGene ID
Hs.371350
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000411.6 NP_000402.3 6, 7 1665
NM_001242784.1 NP_001229713.1 6, 7 2599
NM_001242785.1 NP_001229714.1 6, 7 1758
XM_005260953.3 XP_005261010.1 5, 6 1627
XM_005260955.3 XP_005261012.1 5, 6 1498
XM_005260956.3 XP_005261013.1 5, 6 1674
XM_006723994.2 XP_006724057.1 5, 6 1612
XM_006723995.1 XP_006724058.1 6, 7 1790
XM_011529538.1 XP_011527840.1 6, 7 1645
XM_011529539.2 XP_011527841.1 6, 7 1650
XM_011529540.2 XP_011527842.1 5, 6 1627
XM_011529541.2 XP_011527843.1 5, 6 1501
XM_017028330.1 XP_016883819.1 5, 6 1448
GenBank mRNA AK314189.1 - 5, 6 1532
BC060787.1 - 6, 7 1623 30343493
D23672.2 - 6, 7 1656
D87328.1 - 6, 7 2416
X80160.1 - 5, 6 1516

View Full Product Details