Gene Transcripts

Gene Symbol
RAB43
Entrez Gene ID
339122
Gene Name
RAB43, member RAS oncogene family
Gene Aliases
RAB11B, RAB41
Chromosome Location
Chr. 3: 129087569 - 129122150 on Build GRCh38
UniGene ID
Hs.381132
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001204883.1 NP_001191812.1 3 418
NM_001204884.1 NP_001191813.1 3 406
NM_001204885.1 NP_001191814.1 3 396
NM_001204886.1 NP_001191815.1 3 433
NM_001204887.1 NP_001191816.1 2 684
NM_001204888.1 NP_001191817.1 2 670
NM_198490.2 NP_940892.1 2 684
GenBank mRNA AK128345.1 - 2 480
AK291014.1 - 2 494
AY166852.2 - 2 662
BC062319.1 - 2 520 4873224
BX331971.2 - 2 671
D45418.1 - 2 473
DA008447.1 - 3 406
DA103180.1 - 3 396
DA317502.1 - 3 398
DA339537.1 - 3 433
DA757569.1 - 3 419

Gene Symbol
ISY1-RAB43
Entrez Gene ID
100534599
Gene Name
ISY1-RAB43 readthrough
Gene Aliases
Chromosome Location
Chr. 3: 129087569 - 129161230 on Build GRCh38
UniGene ID
Hs.512661
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001204890.1 NP_001191819.1 12 1271
GenBank mRNA AB032986.1 - 12 1077

View Full Product Details