Gene Transcripts

Gene Symbol
ANKRD33
Entrez Gene ID
341405
Gene Name
ankyrin repeat domain 33
Gene Aliases
C12orf7, PANKY
Chromosome Location
Chr. 12: 51888009 - 51891721 on Build GRCh38
UniGene ID
Hs.433492
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001130015.1 NP_001123487.1 4 717
NM_001304459.1 NP_001291388.1 4 717
NM_001304460.1 NP_001291389.1 3 591
NM_182608.3 NP_872414.3 4 721
XM_017019258.1 XP_016874747.1 4 641
GenBank mRNA AF517115.1 - 1 17
AF517116.1 - 1 17
AK125517.1 - 4 770
BC121152.1 - 3 591
BC121153.2 - 3 950 40121303
BX537908.1 - 4 492
BX640656.1 - 4 492
BX647684.1 - 2 920
BX649168.1 - 4 463

View Full Product Details