Gene Transcripts

Gene Symbol
VSTM2B
Entrez Gene ID
342865
Gene Name
V-set and transmembrane domain containing 2B
Gene Aliases
Chromosome Location
Chr. 19: 29525431 - 29564555 on Build GRCh38
UniGene ID
Hs.451618
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_011526902.2 XP_011525204.1 2 357
XM_011526903.2 XP_011525205.1 2 357

View Full Product Details