Gene Transcripts

Gene Symbol
IL17A
Entrez Gene ID
3605
Gene Name
interleukin 17A
Gene Aliases
CTLA-8, CTLA8, IL-17, IL-17A, IL17
Chromosome Location
Chr. 6: 52186387 - 52190638 on Build GRCh38
UniGene ID
Hs.41724
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002190.2 NP_002181.1 2, 3 267
GenBank mRNA BC066251.1 - 2, 3 265
BC066252.1 - 2, 3 265 6971876
BC066253.1 - 2, 3 265 6971878
BC067503.1 - 2, 3 265 6971877
BC067504.1 - 2, 3 265 6971879
BC067505.1 - 2, 3 265 6971881
HM535222.1 - 1, 2 86
U32659.1 - 2, 3 275
Z58820.1 - 2, 3 264

View Full Product Details