Gene Transcripts

Gene Symbol
KIF5C
Entrez Gene ID
3800
Gene Name
kinesin family member 5C
Gene Aliases
CDCBM2, KINN, NKHC, NKHC-2, NKHC2
Chromosome Location
Chr. 2: 148875223 - 149026759 on Build GRCh38
UniGene ID
Hs.435557
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004522.2 NP_004513.1 20 2641
NR_111932.1 13 1663
XM_011511157.2 XP_011509459.1 13 1722
XM_017004062.1 XP_016859551.1 20 2632
GenBank mRNA AB011103.1 - 20 2614
AB209191.1 - 1 524
BC110287.1 - 13 1593 5165262
BC127772.1 - 17 1965

View Full Product Details