Gene Transcripts

Gene Symbol
TMEM41B
Entrez Gene ID
440026
Gene Name
transmembrane protein 41B
Gene Aliases
Chromosome Location
Chr. 11: 9280654 - 9314768 on Build GRCh38
UniGene ID
Hs.594563
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015012.3 NP_055827.1 7 1115
NR_028491.2 7 1115
GenBank mRNA AK303339.1 - 7 806
BC035034.1 - 7 993 5259179
BC044597.1 - 7 984 4811565
BC064366.1 - 7 872
BC091524.1 - 7 914 30512350
D26067.1 - 7 921

View Full Product Details