Gene Transcripts
- Gene Symbol
- P2RX5
- Entrez Gene ID
- 5026
- Gene Name
- purinergic receptor P2X 5
- Gene Aliases
- LRH-1, P2X5, P2X5R
- Chromosome Location
- Chr. 17: 3673227 - 3696404 on Build GRCh38
- UniGene ID
- Hs.731607
- Species
- Homo sapiens
- Gene Symbol
- P2RX5-TAX1BP3
- Entrez Gene ID
- 100533970
- Gene Name
- P2RX5-TAX1BP3 readthrough (NMD candidate)
- Gene Aliases
- Chromosome Location
- Chr. 17: 3662893 - 3696404 on Build GRCh38
- UniGene ID
- Hs.731607
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_037928.1 |
|
2 |
549 |
|
GenBank mRNA |
AB209623.1 |
- |
2 |
339 |
|
View Full Product Details