Gene Transcripts

Gene Symbol
DHH
Entrez Gene ID
50846
Gene Name
desert hedgehog
Gene Aliases
GDXYM, HHG-3, SRXY7
Chromosome Location
Chr. 12: 49086656 - 49094819 on Build GRCh38
UniGene ID
Hs.524382
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021044.3 NP_066382.1 1, 2 616
GenBank mRNA AK313360.1 - 1, 2 586
BC033507.1 - 1, 2 616 5169286
HQ607677.1 - 1, 2 613
HQ607678.1 - 1, 2 620
U59748.1 - 1, 2 59

View Full Product Details