Gene Transcripts

Gene Symbol
SF3B6
Entrez Gene ID
51639
Gene Name
splicing factor 3b subunit 6
Gene Aliases
CGI-110, HSPC175, Ht006, P14, SAP14, SAP14a, SF3B14, SF3B14a
Chromosome Location
Chr. 2: 24067584 - 24076444 on Build GRCh38
UniGene ID
Hs.177861
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016047.3 NP_057131.1 3 451
GenBank mRNA AF151868.1 - 3 447
AF161523.1 - 3 345
AF184213.1 - 3 447
AF401310.1 - 3 236
AK000621.1 - 3 351
AK311118.1 - 2 238
AK311908.1 - 3 384
BC015463.1 - 3 451 3914184
BG200650.1 - 2 212

View Full Product Details