Gene Transcripts

Gene Symbol
PHIP
Entrez Gene ID
55023
Gene Name
pleckstrin homology domain interacting protein
Gene Aliases
BRWD2, DCAF14, WDR11, ndrp
Chromosome Location
Chr. 6: 78934419 - 79078294 on Build GRCh38
UniGene ID
Hs.511817
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017934.5 NP_060404.3 7, 8 821
XM_005248729.4 XP_005248786.1 7, 8 821
XM_011535918.2 XP_011534220.1 4, 5 477
XM_011535919.1 XP_011534221.1 7, 8 821
XM_017010989.1 XP_016866478.1 8, 9 899
XM_017010990.1 XP_016866479.1 5, 6 497
GenBank mRNA AJ303102.1 - 7, 8 608
AK057039.1 - 7, 8 763
BC021905.1 - 7, 8 710
BC137488.1 - 7, 8 701 9021488
DQ924532.1 - 7, 8 595

View Full Product Details