Gene Transcripts

Gene Symbol
PRCC
Entrez Gene ID
5546
Gene Name
papillary renal cell carcinoma (translocation-associated)
Gene Aliases
RCCP1, TPRC
Chromosome Location
Chr. 1: 156767482 - 156800819 on Build GRCh38
UniGene ID
Hs.516948
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005973.4 NP_005964.3 3 955
XM_005245313.2 XP_005245370.1 3 955
XM_005245314.1 XP_005245371.1 3 955
XM_011509726.1 XP_011508028.1 3 955
GenBank mRNA AK289872.1 - 3 895
BC004913.2 - 3 871 3535413
BC010450.1 - 3 883 4179105
KU178273.1 - 3 665
KU178274.1 - 3 665
KU178275.1 - 3 665
X97124.1 - 3 855

View Full Product Details