Gene Transcripts

Gene Symbol
DDX60
Entrez Gene ID
55601
Gene Name
DEXD/H-box helicase 60
Gene Aliases
Chromosome Location
Chr. 4: 168216291 - 168318807 on Build GRCh38
UniGene ID
Hs.591710
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017631.5 NP_060101.3 5 613
XM_011532103.2 XP_011530405.1 5 613
XM_011532104.2 XP_011530406.1 5 613
XM_017008383.1 XP_016863872.1 5 613
XM_017008384.1 XP_016863873.1 5 613
GenBank mRNA AK307404.1 - 5 613
BC038115.2 - 5 558 5261001

View Full Product Details