Gene Transcripts

Gene Symbol
FAM222B
Entrez Gene ID
55731
Gene Name
family with sequence similarity 222 member B
Gene Aliases
C17orf63
Chromosome Location
Chr. 17: 28755978 - 28855036 on Build GRCh38
UniGene ID
Hs.564533
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001077498.2 NP_001070966.1 3 333
NM_001288631.1 NP_001275560.1 4 504
NM_001288632.1 NP_001275561.1 5 415
NM_001288633.1 NP_001275562.1 4 409
NM_001288634.1 NP_001275563.1 3 402
NM_001288635.1 NP_001275564.1 4 381
NM_018182.3 NP_060652.2 4 367
XM_017024831.1 XP_016880320.1 4 457
XM_017024832.1 XP_016880321.1 3 722
XM_017024833.1 XP_016880322.1 3 262
XM_017024834.1 XP_016880323.1 3 460
GenBank mRNA AK001562.1 - 4 318
AK057665.1 - 3 317
AK095569.1 - 3 402
BC021228.2 - 3 217 4040385
DA185693.1 - 4 320
DA787679.1 - 3 286
DB453514.1 - 5 364
DC374179.1 - 4 297
DC381712.1 - 4 325
HY012898.1 - 4 376
HY040104.1 - 3 324

View Full Product Details