Gene Transcripts

Gene Symbol
CISD1
Entrez Gene ID
55847
Gene Name
CDGSH iron sulfur domain 1
Gene Aliases
C10orf70, MDS029, ZCD1, mitoNEET
Chromosome Location
Chr. 10: 58269102 - 58290553 on Build GRCh38
UniGene ID
Hs.370102
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018464.4 NP_060934.1 2 250
XM_017016409.1 XP_016871898.1 3 491
GenBank mRNA AF220049.1 - 2 189
AK312017.1 - 2 125
AY960578.1 - 2 133
BC005962.1 - 2 226 4051044
BC007043.1 - 2 182 3930701
BC008474.1 - 2 187 4283253
BC059168.1 - 2 197 4132410
DA417986.1 - 2 250

View Full Product Details