Gene Transcripts

Gene Symbol
PRR21
Entrez Gene ID
643905
Gene Name
proline rich 21
Gene Aliases
Chromosome Location
Chr. 2: 240041813 - 240042982 on Build GRCh38
UniGene ID
Hs.693479
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001080835.1 NP_001074304.1 1 292

View Full Product Details