Gene Transcripts

Gene Symbol
LOC101927637
Entrez Gene ID
101927637
Gene Name
uncharacterized LOC101927637
Gene Aliases
Chromosome Location
Chr. 12: 127631269 - 127636467 on Build GRCh38
UniGene ID
Hs.170413
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_120451.1 4 368
GenBank mRNA AI016783.1 - 1 353
BU567943.1 - 3 318

View Full Product Details