Gene Transcripts

Gene Symbol
SLC30A5
Entrez Gene ID
64924
Gene Name
solute carrier family 30 member 5
Gene Aliases
ZNT5, ZNTL1, ZTL1, ZnT-5
Chromosome Location
Chr. 5: 69093949 - 69131072 on Build GRCh38
UniGene ID
Hs.631975
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022902.4 NP_075053.2 1, 2 380
NM_024055.4 NP_076960.1 1, 2 380
XM_006714672.3 XP_006714735.1 1, 2 374
GenBank mRNA AF461760.1 - 1, 2 274
AK292925.1 - 1, 2 244
AY089991.1 - 1, 2 73
AY358505.1 - 1, 2 291
BC000808.1 - 1, 2 225
BC017441.1 - 1, 2 338
BC130452.1 - 1, 2 131
BC130454.1 - 1, 2 131
BX537394.1 - 1, 2 73
DB090660.1 - 1, 2 380
DB259372.1 - 1, 2 374

View Full Product Details