Gene Transcripts

Gene Symbol
TMEM236
Entrez Gene ID
653567
Gene Name
transmembrane protein 236
Gene Aliases
FAM23A, FAM23B, bA162I21.2, bA16O1.2
Chromosome Location
Chr. 10: 17752110 - 17800916 on Build GRCh38
UniGene ID
Hs.564139
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001098844.2 NP_001092314.1 4 927
XM_011519627.2 XP_011517929.1 4 804
XM_011519628.1 XP_011517930.1 3 645
XM_011519629.1 XP_011517931.1 3 693
XM_017016574.1 XP_016872063.1 4 1653
GenBank mRNA AK300456.1 - 3 670
BC157856.1 - 4 927 9057075

View Full Product Details