Gene Transcripts

Gene Symbol
PLEKHA3
Entrez Gene ID
65977
Gene Name
pleckstrin homology domain containing A3
Gene Aliases
FAPP1
Chromosome Location
Chr. 2: 178480410 - 178516324 on Build GRCh38
UniGene ID
Hs.41086
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019091.3 NP_061964.3 1, 2 444
XM_011511689.2 XP_011509991.1 1, 2 506
GenBank mRNA AF286162.1 - 1, 2 283
AK074209.1 - 1, 2 419
AK314459.1 - 1, 2 448
BC035471.1 - 2, 3 298
BC044567.1 - 1, 2 138 5262097
BM794932.1 - 1, 2 444

View Full Product Details