Gene Transcripts
- Gene Symbol
- LINC01346
- Entrez Gene ID
- 728716
- Gene Name
- long intergenic non-protein coding RNA 1346
- Gene Aliases
- Chromosome Location
- Chr. 1: 3940612 - 3952583 on Build GRCh38
- UniGene ID
- Hs.668359
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_040065.1 |
|
2 |
209 |
|
GenBank mRNA |
AK124708.1 |
- |
2 |
204 |
|
DB288420.1 |
- |
2 |
256 |
|
View Full Product Details