Gene Transcripts

Gene Symbol
C6
Entrez Gene ID
729
Gene Name
complement component 6
Gene Aliases
Chromosome Location
Chr. 5: 41142116 - 41261486 on Build GRCh38
UniGene ID
Hs.481992
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000065.3 NP_000056.2 16 2583
NM_001115131.2 NP_001108603.2 16 2610
XM_005248357.2 XP_005248414.1 16 2658
XM_006714496.3 XP_006714559.1 17 2715
XM_011514114.2 XP_011512416.1 17 2715
XM_011514115.2 XP_011512417.1 17 2636
XM_011514116.2 XP_011512418.1 18 2752
XM_011514117.2 XP_011512419.1 18 2794
XM_011514118.2 XP_011512420.1 17 2532
XM_011514119.2 XP_011512421.1 16 2658
XM_011514121.2 XP_011512423.1 12 1801
XM_017009818.1 XP_016865307.1 18 2775
XM_017009819.1 XP_016865308.1 13 1978
GenBank mRNA AB126592.1 - 15 2345
AK292509.1 - 16 2481
AK313673.1 - 16 2502
BC035723.1 - 16 2610 5591446
J05024.1 - 16 2402
J05064.1 - 16 2500

View Full Product Details