Gene Transcripts

Gene Symbol
VCL
Entrez Gene ID
7414
Gene Name
vinculin
Gene Aliases
CMD1W, CMH15, HEL114, MV, MVCL
Chromosome Location
Chr. 10: 73998114 - 74120160 on Build GRCh38
UniGene ID
Hs.643896
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003373.3 NP_003364.1 14 2045
NM_014000.2 NP_054706.1 14 2045
GenBank mRNA AK126978.1 - 14 2300
AK304823.1 - 13 1974
AL832130.1 - 7 967
BC039174.1 - 14 2032 4520338
BX537994.1 - 16 4755
EU794673.1 - 14 2045
M33308.1 - 14 2001

View Full Product Details