Gene Transcripts

Gene Symbol
XPNPEP2
Entrez Gene ID
7512
Gene Name
X-prolyl aminopeptidase 2
Gene Aliases
AEACEI, APP2
Chromosome Location
Chr. X: 129738970 - 129769549 on Build GRCh38
UniGene ID
Hs.170499
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003399.5 NP_003390.4 8 903
GenBank mRNA BC126174.1 - 8 820
BC143901.1 - 8 820 9052419
BF830085.1 - 2 148
U90724.2 - 8 923

View Full Product Details