Gene Transcripts

Gene Symbol
CHAC1
Entrez Gene ID
79094
Gene Name
ChaC glutathione specific gamma-glutamylcyclotransferase 1
Gene Aliases
Chromosome Location
Chr. 15: 40942135 - 40956519 on Build GRCh38
UniGene ID
Hs.155569
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024111.3 NP_077016.2 3 548
XM_017022563.1 XP_016878052.1 4 5072
XM_017022564.1 XP_016878053.1 5 2854
XM_017022565.1 XP_016878054.1 4 5061
GenBank mRNA BC001683.1 - 3 502
BC001847.2 - 3 493 2964714
BC019625.1 - 3 504 4473135
BQ900544.1 - 3 548

View Full Product Details