Gene Transcripts

Gene Symbol
ALG13
Entrez Gene ID
79868
Gene Name
ALG13, UDP-N-acetylglucosaminyltransferase subunit
Gene Aliases
CDG1S, CXorf45, EIEE36, GLT28D1, MDS031, TDRD13, YGL047W
Chromosome Location
Chr. X: 111680741 - 111764164 on Build GRCh38
UniGene ID
Hs.443061
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099922.2 NP_001093392.1 2, 3 352
NM_001168385.2 NP_001161857.1 2, 3 352
NM_001257230.1 NP_001244159.1 2, 3 248
NM_001257234.1 NP_001244163.1 2, 3 321
NM_001257235.2 NP_001244164.1 2, 3 321
NM_001257237.1 NP_001244166.1 2, 3 425
NM_001257239.2 NP_001244168.1 2, 3 340
NM_001257240.2 NP_001244169.1 2, 3 425
NM_001324291.1 NP_001311220.1 2, 3 248
NM_001324292.1 NP_001311221.1 2, 3 352
NM_001324293.1 NP_001311222.1 2, 3 425
NM_001324294.1 NP_001311223.1 2, 3 236
NM_018466.5 NP_060936.1 2, 3 352
NR_033125.2 2, 3 248
XM_006724693.2 XP_006724756.1 2, 3 725
XM_006724697.3 XP_006724760.2 2, 3 625
XM_006724698.3 XP_006724761.2 2, 3 750
XM_011531028.2 XP_011529330.1 2, 3 726
XM_011531030.1 XP_011529332.1 2, 3 724
XM_011531031.1 XP_011529333.1 2, 3 425
XM_011531032.1 XP_011529334.1 2, 3 231
XM_011531033.2 XP_011529335.1 2, 3 271
XM_017029846.1 XP_016885335.1 2, 3 727
XM_017029847.1 XP_016885336.1 2, 3 726
XM_017029848.1 XP_016885337.1 2, 3 724
XM_017029849.1 XP_016885338.1 2, 3 725
XM_017029850.1 XP_016885339.1 2, 3 724
XM_017029851.1 XP_016885340.1 2, 3 727
GenBank mRNA AF220051.1 - 2, 3 283
AK300394.1 - 2, 3 190
AK302729.1 - 2, 3 293
AK304712.1 - 2, 3 242
AK312229.1 - 2, 3 291
AK316306.1 - 2, 3 294
AK316522.1 - 2, 3 369
AL524204.3 - 2, 3 171
BC005336.1 - 2, 3 285 3996922
BE747995.1 - 2, 3 281
BF699152.1 - 2, 3 353
BG621042.1 - 1, 2 135
BM759764.1 - 2, 3 334
CB268871.1 - 2, 3 299
DA830058.1 - 2, 3 416
DB292609.1 - 2, 3 425
DC400909.1 - 2, 3 236
DN992019.1 - 2, 3 420

View Full Product Details