Gene Transcripts

Gene Symbol
C10orf95
Entrez Gene ID
79946
Gene Name
chromosome 10 open reading frame 95
Gene Aliases
Chromosome Location
Chr. 10: 102449837 - 102451543 on Build GRCh38
UniGene ID
Hs.225084
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024886.2 NP_079162.1 2 214
GenBank mRNA AK024342.1 - 2 214
BC126459.1 - 2 159 8992175

View Full Product Details