Gene Transcripts

Gene Symbol
ZFP2
Entrez Gene ID
80108
Gene Name
ZFP2 zinc finger protein
Gene Aliases
ZNF751, zfp-2
Chromosome Location
Chr. 5: 178895894 - 178933212 on Build GRCh38
UniGene ID
Hs.654533
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030613.3 NP_085116.2 5 631
GenBank mRNA AK131367.1 - 5 610
BC132779.1 - 3 404 40146801
BC132781.1 - 3 429 40146803
BC142989.1 - 5 600 8860436
BC144213.1 - 3 404 9052733
BC144216.1 - 4 515 9052736

View Full Product Details