Gene Transcripts

Gene Symbol
AMN
Entrez Gene ID
81693
Gene Name
amnion associated transmembrane protein
Gene Aliases
PRO1028, amnionless
Chromosome Location
Chr. 14: 102922430 - 102930842 on Build GRCh38
UniGene ID
Hs.534494
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030943.3 NP_112205.2 2 128
XM_011537202.2 XP_011535504.1 2 373
XM_011537203.2 XP_011535505.1 2 629
GenBank mRNA AF328788.1 - 2 117
AK055802.1 - 1 841
AY358468.1 - 2 115
BC029948.1 - 2 128

View Full Product Details