Gene Transcripts
- Gene Symbol
- MADCAM1
- Entrez Gene ID
- 8174
- Gene Name
- mucosal vascular addressin cell adhesion molecule 1
- Gene Aliases
- MACAM1
- Chromosome Location
- Chr. 19: 496490 - 505343 on Build GRCh38
- UniGene ID
- Hs.102598
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_130760.2 |
NP_570116.2
|
2, 3 |
337 |
|
NM_130762.2 |
NP_570118.1
|
2, 3 |
337 |
|
GenBank mRNA |
BC136408.1 |
- |
2, 3 |
373 |
|
BC137506.1 |
- |
2, 3 |
373 |
9021506 |
BC137507.1 |
- |
2, 3 |
373 |
9021507 |
BC142629.1 |
- |
2, 3 |
333 |
40147551 |
BC144065.1 |
- |
2, 3 |
373 |
9052585 |
DQ076657.1 |
- |
2, 3 |
327 |
|
U43628.1 |
- |
2, 3 |
327 |
|
U82483.1 |
- |
2, 3 |
337 |
|
View Full Product Details